Learn More
Sigma Aldrich Fine Chemicals Biosciences CelLyticTM Express
Catalog No. 502905827
Encompass
CelLytic(TM) Express is a non-denaturing and highly efficient protein extraction formulation for in-culture bacterial cell lysis. This proprietary powder formulation extracts 2-3 times more protein than conventional methods such as sonication. In addition the method is fast and requires less sample manipulations reducing proteolytic degradation and preserving protein activity. Unlike other in-culture lysis products CelLytic(TM) Express is a complete formulation including lysozyme and endonuclease. The resulting lysate is completely clear of cellular debris and is immediately ready for affinity purification. CelLytic(TM) Express unique powder formulation adds minimal volume to the final lysate. It provides a cell lysis method with defined volume and time parameters making it easier to validate and more convenient for production scale protocols.
Catalog No. 50-290-5827 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C199010X25ML
Add to Cart
Add to Cart
CelLytic(TM) Express is a non-denaturing and highly efficient protein extraction formulation for in-culture bacterial cell lysis. This proprietary powder formulation extracts 2-3 times more protein than conventional methods such as sonication. In addition the method is fast and requires less sample manipulations reducing proteolytic degradation and preserving protein activity. Unlike other in-culture lysis products CelLytic(TM) Express is a complete formulation including lysozyme and endonuclease. The resulting lysate is completely clear of cellular debris and is immediately ready for affinity purification. CelLytic(TM) Express unique powder formulation adds minimal volume to the final lysate. It provides a cell lysis method with defined volume and time parameters making it easier to validate and more convenient for production scale protocols.