Learn More
Sigma Aldrich Fine Chemicals Biosciences CelLyticTM NuCLEARTM Extra
Catalog No. 502904472
Encompass
The procedure for the nuclear protein extraction method is to allow cells to swell with hypotonic buffer. The cells are then disrupted the cytoplasmic fraction is removed and the nuclear proteins are released from the nuclei by a high salt buffer.
Catalog No. 50-290-4472 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NXTRACT1KT
The procedure for the nuclear protein extraction method is to allow cells to swell with hypotonic buffer. The cells are then disrupted the cytoplasmic fraction is removed and the nuclear proteins are released from the nuclei by a high salt buffer.