Learn More
Sigma Aldrich Fine Chemicals Biosciences CelLyticTM PN Isolation/Ex
Catalog No. 502905221
Encompass
CelLytic(TM)-PN extraction kit is for the isolation of nuclei and extraction of functional nuclear proteins from plant leaves. Nuclei or nuclear proteins can be extracted from few grams to hundreds grams of fresh and frozen leaves. The nuclear protein extract is suitable for the detection of DNA-protein interactions using gel-shift assay DNase-I footprinting analysis as well as western blot assay and similar techniques. The isolated nuclei can also be used as a source for chromatin genomic DNA and histones.
Catalog No. 50-290-5221 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CELLYTPN11KT
CelLytic(TM)-PN extraction kit is for the isolation of nuclei and extraction of functional nuclear proteins from plant leaves. Nuclei or nuclear proteins can be extracted from few grams to hundreds grams of fresh and frozen leaves. The nuclear protein extract is suitable for the detection of DNA-protein interactions using gel-shift assay DNase-I footprinting analysis as well as western blot assay and similar techniques. The isolated nuclei can also be used as a source for chromatin genomic DNA and histones.