Learn More
Sigma Aldrich Fine Chemicals Biosciences CelLyticTM Y Cell Lysis Re
Catalog No. 502901635
Encompass
CelLytic Y Cell Lysis Reagent is versatile phosphate-free and non-denaturing. It provides an effective method for cell lysis and protein solubilization. Target proteins maintain immunoreactivity or biological function. The protocol is brief and preformed at room temperature. No extreme conditions or glass beads are required. Use only 2.5-5 ml/gram yeast cells.
Catalog No. 50-290-1635 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C448250ML
CelLytic Y Cell Lysis Reagent is versatile phosphate-free and non-denaturing. It provides an effective method for cell lysis and protein solubilization. Target proteins maintain immunoreactivity or biological function. The protocol is brief and preformed at room temperature. No extreme conditions or glass beads are required. Use only 2.5-5 ml/gram yeast cells.