Learn More
Celprogen CELPROGEN Human Bone Marrow Derived Endothelial Stem Cell Complete Media, Serum, 500 mL
Catalog No. NC3971777
Encompass
This product is Human Bone Marrow Derived Endothelial Stem Cell Complete Media with Serum.
  • Tissue tested, including stem cells.
  • Supplied as a 500 mL sterile filtered unit.
  • Available as a pack of 6, 500 mL unit sizes.
  • Also available without serum.
Catalog No. NC3971777 Supplier Celprogen Supplier No. M3609626S
Add to Cart
Add to Cart
This product is Human Bone Marrow Derived Endothelial Stem Cell Complete Media with Serum.
  • Tissue tested, including stem cells.
  • Supplied as a 500 mL sterile filtered unit.
  • Available as a pack of 6, 500 mL unit sizes.
  • Also available without serum.