Learn More
Lifeline Cell Technology CELS FIBS HU AD DRML LIFELN VL
SDP
Catalog No. 50311859
Encompass_Preferred
Lifeline normal Human Dermal Fibroblasts adult are cryopreserved as primary cells. Quality tested in both FibroLife Serum-Free Medium and FibroLife S2 Medium to ensure optimal growth.
Catalog No. 50-311-859 Supplier Lifeline Cell Technology Supplier No. FC0024
Lifeline normal Human Dermal Fibroblasts adult are cryopreserved as primary cells. Quality tested in both FibroLife Serum-Free Medium and FibroLife S2 Medium to ensure optimal growth.