Learn More
Lifeline Cell Technology CELS MELS HU NEO P1 LIFELNE VL
SDP
Catalog No. 50311858
Encompass_Preferred
Normal Human Epidermal Melanocytes neonatal are cryopreserved as primary or secondary cells. Quality tested in DermaLife M Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.
Catalog No. 50-311-858 Supplier Lifeline Cell Technology Supplier No. FC0023
Normal Human Epidermal Melanocytes neonatal are cryopreserved as primary or secondary cells. Quality tested in DermaLife M Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.