Learn More
Lifeline Cell Technology CELS MELS HU NEO P2 LIFELNE VL
SDP
Catalog No. 50311856
Encompass_Preferred
Normal Human Epidermal Melanocytes neonatal are cryopreserved as primary or secondary cells. Quality tested in DermaLife M Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.
Catalog No. 50-311-856 Supplier Lifeline Cell Technology Supplier No. FC0019
Add to Cart
Add to Cart
Normal Human Epidermal Melanocytes neonatal are cryopreserved as primary or secondary cells. Quality tested in DermaLife M Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.