Learn More
Lifeline Cell Technology CELS RENAL EPI MX HU LIFELN VL
SDP
Catalog No. 50311854
Encompass_Preferred
Normal Human Renal Mixed Epithelial Cells are cryopreserved as primary cells. Quality tested in RenaLife Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.
Catalog No. 50-311-854 Supplier Lifeline Cell Technology Supplier No. FC0017
Normal Human Renal Mixed Epithelial Cells are cryopreserved as primary cells. Quality tested in RenaLife Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.