Learn More
Lifeline Cell Technology CELS RENAL PT EPI HU LIFELN VL
SDP
Catalog No. 50311850
Encompass_Preferred
Normal Human Renal Proximal Tubule Epithelial Cells are cryopreserved as secondary cells. Quality tested in RenaLife Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.
Catalog No. 50-311-850 Supplier Lifeline Cell Technology Supplier No. FC0013
Normal Human Renal Proximal Tubule Epithelial Cells are cryopreserved as secondary cells. Quality tested in RenaLife Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.