Learn More
ACROBiosystems CelThera GMP T Cell Expansion Medium, 1 L
Catalog No. NC3864341
Encompass_Preferred
CelThera™ GMP T Cell Expansion Medium is a GMP-grade cell culture medium formulated to support robust expansion of T cells for clinical and manufacturing workflows. Supplied in a 1 L format, the medium is intended for integration into GMP-compliant processes where consistent performance is required.
  • GMP-grade formulation for regulatory-compliant workflows.
  • Optimized to support T cell expansion.
  • Supplied as 1 L liquid suitable for process development and small-scale production.
  • Intended for use in cell therapy manufacturing and related applications.
Catalog No. NC3864341 Supplier ACROBiosystems Supplier No. GMPCM31031L
CelThera™ GMP T Cell Expansion Medium is a GMP-grade cell culture medium formulated to support robust expansion of T cells for clinical and manufacturing workflows. Supplied in a 1 L format, the medium is intended for integration into GMP-compliant processes where consistent performance is required.
  • GMP-grade formulation for regulatory-compliant workflows.
  • Optimized to support T cell expansion.
  • Supplied as 1 L liquid suitable for process development and small-scale production.
  • Intended for use in cell therapy manufacturing and related applications.