Learn More
Cayman Chemical CelularMetabolIsmScreeng50uL
SDP
Catalog No. 502949750
Encompass_Preferred
A GABAB receptor agonist increases latency to paw or tail withdrawal in the hot-plate test in mice at 1 and 3 mg/kg reduces the number of lever responses for alcohol in Sardinian alcohol-preferring rats at 3 mg/kg
Catalog No. 50-294-9750 Supplier Cayman Chemical Supplier No. 3370550 UL
A GABAB receptor agonist increases latency to paw or tail withdrawal in the hot-plate test in mice at 1 and 3 mg/kg reduces the number of lever responses for alcohol in Sardinian alcohol-preferring rats at 3 mg/kg