Learn More
Premier Lab Supply CEMBLES Pre-Assembled XRF Sample Cups – PN# PA-240-35-160-LS
SDP
Catalog No. NC1731205
Encompass_Preferred
CEMBLES Pre-Assembled XRF Sample Cups! (CEMBLES PN# PA-240-35-160-LS; 35mm Cups PN# SC-3335; Mylar Low Sulfur film 6.0μm; 48 per pack). CEMBLES are custom made to your specifications & available in variant configurations of cups and thin films that have been pre-assembled for your convenience.
Catalog No. NC1731205 Supplier Premier Lab Supply Supplier No. PA24035160LS
CEMBLES Pre-Assembled XRF Sample Cups! (CEMBLES PN# PA-240-35-160-LS; 35mm Cups PN# SC-3335; Mylar Low Sulfur film 6.0μm; 48 per pack). CEMBLES are custom made to your specifications & available in variant configurations of cups and thin films that have been pre-assembled for your convenience.