Learn More
Premier Lab Supply CEMBLES Pre-Assembled XRF Sample Cups – PN# PA-280-35-260
SDP
Catalog No. NC1198105
Encompass_Preferred
CEMBLES Pre-Assembled XRF Sample Cups! (CEMBLES PN# PA-280-35-260; 35mm Cups PN# SC-3335; Polypropylene film 6.0μm; 48 per pack). CEMBLES are custom made to your specifications & available in variant configurations of cups and thin films that have been pre-assembled for your convenience. 
Catalog No. NC1198105 Supplier Premier Lab Supply Supplier No. PA28035260
CEMBLES Pre-Assembled XRF Sample Cups! (CEMBLES PN# PA-280-35-260; 35mm Cups PN# SC-3335; Polypropylene film 6.0μm; 48 per pack). CEMBLES are custom made to your specifications & available in variant configurations of cups and thin films that have been pre-assembled for your convenience. 