Learn More
Cayman Chemical β-Cembrenediol, Purity: ≥98%, CAS: 57605-81-9, 10 mg
SDP
Catalog No. 502984237
Encompass_Preferred
A diterpenoid from tobacco plants; inhibits induction of EA-EBV by TPA in vitro (IC50 = 21.9 μM); inhibits tumor promoting effects of TPA on 7,12-demethylbenz[a]anthracene-induced papilloma formation in vivo; exerts autotoxicity and phytotoxic activity against L. sativa seedlings
Catalog No. 50-298-4237 Supplier Cayman Chemical Supplier No. 2103110 MG
A diterpenoid from tobacco plants; inhibits induction of EA-EBV by TPA in vitro (IC50 = 21.9 μM); inhibits tumor promoting effects of TPA on 7,12-demethylbenz[a]anthracene-induced papilloma formation in vivo; exerts autotoxicity and phytotoxic activity against L. sativa seedlings