Learn More
Med Vet International Bone Cement - Medium Viscosity - 20gm, J0912A, Each
Catalog No. 5000495062
Encompass_Preferred
Type 1: Medium Viscosity forms a digitally (finger) malleable putty. Applications: • Cement the acetabular component of a hip replacement • Adjunct to acetabular fracture with pins and screws • Spinal fractures • Management of cervical wobblers by distraction fusion • Filling dead space in equine tooth extractions • Manufacturing of antibiotic impregnated beads (AIPMMA) • Choose to add one's choice of suitable antibiotic or pre-added gentamycin, creating a slow release focal antibiosis. Especially useful in the management of refractory rabbit abscesses. Instructions included.
Catalog No. 50-004-95062 Supplier Med Vet International Supplier No. JOR912A
Type 1: Medium Viscosity forms a digitally (finger) malleable putty. Applications: • Cement the acetabular component of a hip replacement • Adjunct to acetabular fracture with pins and screws • Spinal fractures • Management of cervical wobblers by distraction fusion • Filling dead space in equine tooth extractions • Manufacturing of antibiotic impregnated beads (AIPMMA) • Choose to add one's choice of suitable antibiotic or pre-added gentamycin, creating a slow release focal antibiosis. Especially useful in the management of refractory rabbit abscesses. Instructions included.