Learn More
Medchemexpress LLC HY-111445 5mg Medchemexpress, CeMMEC1 CAS:440662-09-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1114455MG
Encompass_Preferred
Medchemexpress, HY-111445 5mg CeMMEC1 CAS:440662-09-9 CeMMEC1 is an inhibitor of BRD4, and also has high affinity for TAF1, with an IC50 of 0.9 μM for TAF1, and a Kd of 1.8 μM for TAF1 (2). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8870