Learn More
Medchemexpress LLC HY-101088 5mg Medchemexpress, CeMMEC13 CAS:1790895-25-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1010885MG
Encompass_Preferred
Medchemexpress, HY-101088 5mg CeMMEC13 CAS:1790895-25-8 CeMMEC13 is a potent inhibitor of TAF1 (2) bromodomain, with an IC 50 of 2.1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2032