Learn More
Medchemexpress LLC Cemsidomide | 2504235-67-8 | 99.5% | 469.53 g/mol | C28H27N3O4 | 5 MG
SDP
Catalog No. 5000221170
Encompass_Preferred
Cemsidomide is an investigational small-molecule molecular glue that promotes ubiquitin ligase-mediated degradation of the IKZF1 and IKZF3 transcription factors. It is provided for research use in biochemical and cellular studies and is supplied as a solid and as a 10 mM solution in DMSO. Reported molecular formula C28H27N3O4 and molecular weight 469.53 g/mol; reported purity ~99.54%.
  • Promotes IKZF1 and IKZF3 degradation in cellular assays.
  • Available as solid or as a 10 mM solution in DMSO for experimental flexibility.
  • High reported purity suitable for biochemical and cellular studies.
  • Applicable to research on ubiquitin ligase-mediated protein degradation.
  • Offered in multiple small-scale package sizes for laboratory use.
Catalog No. 50-002-21170 Supplier Medchemexpress LLC Supplier No. HY1448415MG
Cemsidomide is an investigational small-molecule molecular glue that promotes ubiquitin ligase-mediated degradation of the IKZF1 and IKZF3 transcription factors. It is provided for research use in biochemical and cellular studies and is supplied as a solid and as a 10 mM solution in DMSO. Reported molecular formula C28H27N3O4 and molecular weight 469.53 g/mol; reported purity ~99.54%.
  • Promotes IKZF1 and IKZF3 degradation in cellular assays.
  • Available as solid or as a 10 mM solution in DMSO for experimental flexibility.
  • High reported purity suitable for biochemical and cellular studies.
  • Applicable to research on ubiquitin ligase-mediated protein degradation.
  • Offered in multiple small-scale package sizes for laboratory use.