Learn More
Cenmed™ Air Filtration System
SDP
Catalog No. 21520116
Change view
Click to view available options
Includes:
Charcoal Filter
Catalog No. Includes
21-520-116 Charcoal Filter
1 options
Catalog No. 21-520-116 Supplier Cenmed™ Supplier No. CEN0030
  • $5,779.00
    $3,185.65 / Each of 1
    Save $2,593.35
    45% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Designed to help minimize your employees' exposure to fumes and odors generated from chemicals caused by the everyday use of solvents, such as alcohol, xylene, toluene, formalin, and many more.
  • Improves air quality in the lab
  • Helps to minimize hazardous fumes to the employees
  • Works quietly and efficiently 7 days a week, 24 hours a day
  • Aids with respect to compliance issues
  • Measures 23 in. high and 11 in. wide
Specifications
Product Type Air Filtration System
Description Compact Air Filtration System
For Use With (Application) Cleanses the air of irritating and potentially harmful vapors
Includes Charcoal Filter