Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cenmed™ Black Biohazard Specimen Bag, 6 x 9 in.
SDP

Catalog No. 21520114
Change view
Click to view available options
Length (English):
6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
21-520-114 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-520-114 Supplier Cenmed™ Supplier No. 187609
  • $576.83
    $492.65 / Case of 1000

    Save $84.18

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Three Wall Medical Transport Specimen Zip Bag With Biohazard Logo

  • 6 × 9 in.
  • A black opaque back keeps patient or confidential information private
  • With a printed biohazard logo, a separate outside pocket holds the requisition
  • No glove-to-adhesive contact, ensuring ease of use
  • Available in single-pocket versions when requisitions are not used, or in diagnostic kit packaging

Specifications

Color Black
Length (English) 6 in.
Width (English) 9 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.