Learn More
Cenmed™ Sensitive Urine & Serum hCG Pregnancy Test Kit
SDP
Catalog No. 21520106
Change view
Click to view available options
Quantity:
25 Tests
Catalog No. Quantity
21-520-106 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. 21-520-106 Supplier Cenmed™ Supplier No. 4230020
Add to Cart
Add to Cart
This item is not returnable. View return policy
Easy and convenient one-step procedure. The Cenmed™ Elite Plus™ hCG One Step Urine/Serum Pregnancy Test is a rapid chromatographic immunoassay for the visual, qualitative detection of human chorionic gonadotropin (hCG) in serum or urine specimen to help in the early determination of pregnancy.
  • Test kits are for health care professional use including professionals at physician's office labs (POLs)
  • Capable of detecing pregnancy as early as six to eight days after implantation, well before the first missed menses
  • Sensitivity of 20 mlU/mL
  • Results in three minutes for urine and five minutes for serum
  • Stable endpoint
  • Easy Patient I.D. labeling system
  • CLIA waived for urine testing
Specifications
Sample Type Serum, Urine
Type Pregnancy Test Kit
Quantity 25 Tests
Test Time 3 min. for urine, 5 min. for serum