Learn More
Cenmed™ Fine Needle Aspiration Kit
SDP
Catalog No. 21520117
Change view
Click to view available options
Product Type:
Fine Needle Aspiration Kit
Catalog No. Product Type
21-520-117 Fine Needle Aspiration Kit
1 options
Catalog No. 21-520-117 Supplier Cenmed™ Supplier No. FNA001
Add to Cart
Add to Cart
The Fine Needle Aspiration Kit provides a customizable solution to optimize your sample preparation.
Kit Includes:
  • Reagent Alcohol 95% Solution (pre-filled fixative inside a coplin jar) 
  • Buffered 10% Neutral Buffered Formalin
  • Microscope slides
  • Slide mailers
  • Patient Label
  • Biohazard transport bag 
  • 15mL centrifuge tubes
Specifications
Product Type Fine Needle Aspiration Kit