Learn More
Cenmed™ Parasitology Stool Transport Kit
SDP
Catalog No. 21520112
Change view
Click to view available options
Includes:
Each kit contains 1 vial of 10% Formalin Fixative and 1 vial of Zn-PVA Fixative. These kits contain NO Mercury.
Catalog No. Includes
21-520-112 Each kit contains 1 vial of 10% Formalin Fixative and 1 vial of Zn-PVA Fixative. These kits contain NO Mercury.
1 options
Catalog No. 21-520-112 Supplier Cenmed™ Supplier No. 3202301
Add to Cart
Add to Cart
Includes
  • 10% Formalin Fixative - 1 vial
  • Zn-PVA Fixative- 1 vial
Vials are used in the routine collection, preservation, and transportation, of stool specimens
  • Broad range with long shelf life
  • Industry standard color coded caps and labels
  • Easy to use
  • “Spork” cap for safe collection
  • Convenient packaging, including bulk kits
  • Multilingual patient collection instructions
Specifications
Includes Each kit contains 1 vial of 10% Formalin Fixative and 1 vial of Zn-PVA Fixative. These kits contain NO Mercury.