Learn More
Aviva Systems Biology Corp CEND1 Antibody - N-terminal region (ARP47273_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47273_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CEND1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CEND1. Alternate names: BM88, FLJ90066, MGC34326ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-59504