Learn More
Medchemexpress LLC HY-13072 5mg Medchemexpress, Cenisertib CAS:871357-89-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY130725MG
Encompass_Preferred
Medchemexpress, HY-13072 5mg Cenisertib CAS:871357-89-0 Cenisertib (AS-703569) is a multi-kinase inhibitor that blocks the activity of Aurora-kinase-A/B, ABL1, AKT, STAT5 and FLT3. Cenisertib induces major growth-inhibitory effects by blocking the activity of several different molecular targets in neoplastic mast cells (MC). Cenisertib inhibits tumor growth in xenograft models of pancreatic, breast, colon, ovarian, and lung tumors and leukemia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9834