Learn More
Medchemexpress LLC Cenisertib | 871357-89-0 | 99.9% | 100 MG
SDP
Catalog No. 5000341037
Encompass_Preferred
Cenisertib (Standard) is an analytical standard of Cenisertib, intended for research and analytical applications. It is an ATP-competitive multi-kinase inhibitor that blocks the activity of Aurora-kinase-A/B, ABL1, AKT, STAT5, and FLT3. It induces significant growth-inhibitory effects by blocking various molecular targets in neoplastic mast cells and inhibits tumor growth in xenograft models of pancreatic, breast, colon, ovarian, and lung tumors and leukemia.
  • ATP-competitive multi-kinase inhibitor
  • Targets Aurora-kinase-A/B, ABL1, AKT, STAT5, and FLT3
  • Induces growth-inhibitory effects in neoplastic mast cells
  • Inhibits tumor growth in xenograft models of various cancers
  • Used as a reference standard in assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS
Catalog No. 50-003-41037 Supplier Medchemexpress LLC Supplier No. HY13072100MG
Cenisertib (Standard) is an analytical standard of Cenisertib, intended for research and analytical applications. It is an ATP-competitive multi-kinase inhibitor that blocks the activity of Aurora-kinase-A/B, ABL1, AKT, STAT5, and FLT3. It induces significant growth-inhibitory effects by blocking various molecular targets in neoplastic mast cells and inhibits tumor growth in xenograft models of pancreatic, breast, colon, ovarian, and lung tumors and leukemia.
  • ATP-competitive multi-kinase inhibitor
  • Targets Aurora-kinase-A/B, ABL1, AKT, STAT5, and FLT3
  • Induces growth-inhibitory effects in neoplastic mast cells
  • Inhibits tumor growth in xenograft models of various cancers
  • Used as a reference standard in assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS