Learn More
Medchemexpress LLC Cenisertib | 871357-89-0 | 99.9% | C24H30FN7O | 10MG
SDP
Catalog No. 5000259000
Encompass_Preferred
Cenisertib is an ATP-competitive multi-kinase inhibitor used in preclinical research to block Aurora kinases and other oncogenic signaling pathways. It is provided as a high-purity analytical standard or reagent in solid and solution forms for cellular, biochemical, and xenograft studies.
  • ATP-competitive multi-kinase inhibitor targeting Aurora A/B, ABL1, AKT, STAT5, and FLT3.
  • High purity (99.9%) suitable for analytical and biological assays.
  • Available as solid and as 10 mM solution in DMSO for flexible use.
  • Demonstrated antitumor activity in multiple xenograft models.
  • Recommended storage conditions for powder and solvent to preserve stability.
Catalog No. 50-002-59000 Supplier Medchemexpress LLC Supplier No. HY1307210MG
Cenisertib is an ATP-competitive multi-kinase inhibitor used in preclinical research to block Aurora kinases and other oncogenic signaling pathways. It is provided as a high-purity analytical standard or reagent in solid and solution forms for cellular, biochemical, and xenograft studies.
  • ATP-competitive multi-kinase inhibitor targeting Aurora A/B, ABL1, AKT, STAT5, and FLT3.
  • High purity (99.9%) suitable for analytical and biological assays.
  • Available as solid and as 10 mM solution in DMSO for flexible use.
  • Demonstrated antitumor activity in multiple xenograft models.
  • Recommended storage conditions for powder and solvent to preserve stability.