Learn More
Enzo Life Sciences CENP A monoclonal antibody (3-19) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCC006E
Encompass_Preferred
Clone: 3-19. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of human CENP-A. UniProt: P49450. Species Reactivity: Human. Applications: ICC, IHC, WB. Recommended dilutions: Immunocytochemistry (10µg/ml). Immunohistochemistry (5µg/ml). Western Blot (1µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-666-433