Learn More
Aviva Systems Biology Corp CENPA Antibody - N-terminal region (ARP46040_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46040_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CENPA. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CENPA. Alternate names: CenH3, CENP-AApplicationIHC, WB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-59192