Learn More
Aviva Systems Biology Corp CENPA Antibody - middle region (ARP46041_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46041_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CENPA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CENPA. Alternate names: CenH3, CENP-AApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59193