Learn More
Aviva Systems Biology Corp CENPB Antibody - C-terminal region (ARP51969_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51969_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CENPB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CENPB. Alternate names: -ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-60659