Learn More
Aviva Systems Biology Corp CENPH Antibody - N-terminal region (ARP58265_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58265_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CENPH. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CENPH. Alternate names: NNF1, PMF1ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-286-459