Learn More
Aviva Systems Biology Corp CENPH Antibody - middle region (ARP58266_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58266_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CENPH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CENPH. Alternate names: NNF1, PMF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54078