Learn More
Aviva Systems Biology Corp CENPM Antibody - middle region (ARP46323_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46323_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CENPM. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CENPM. Alternate names: C22orf18, CENP-M, MGC861, PANE1, bK250D10.2ApplicationIF, WB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-59302