Learn More
Beckman Coulter Polycarbonate Bottle with Screw-On Cap, 62 x 120mm, 250 mL, 6/pk
Catalog No. NC9312712
Encompass_Preferred
Polycarbonate wide mouth bottle with cap assemblies for use in high performance and high capacity rotors.
Catalog No. NC9312712 Supplier Beckman Coulter Supplier No. 356013
Polycarbonate wide mouth bottle with cap assemblies for use in high performance and high capacity rotors.