Learn More
Chemglass Life Sciences Bottle, Centrifuge, 250mL, Heavy Wall, Rubber Stopper # 6
SDP
Catalog No. NC1588966
Encompass_Preferred
Bottle, Centrifuge, 250mL, Heavy Wall, Rubber Stopper # 6. For use where relatively small amounts of solids are involved. These bottles have a small bottom area, which permits concentration of sediment for decanting operations. Due to the heavy wall, the actual capacity is approximately 85% of the capacity listed. The 250mL fits IEC #384, the 500mL fits IEC #353.
Catalog No. NC1588966 Supplier Chemglass Life Sciences Supplier No. CG8066250
Bottle, Centrifuge, 250mL, Heavy Wall, Rubber Stopper # 6. For use where relatively small amounts of solids are involved. These bottles have a small bottom area, which permits concentration of sediment for decanting operations. Due to the heavy wall, the actual capacity is approximately 85% of the capacity listed. The 250mL fits IEC #384, the 500mL fits IEC #353.