Learn More
Andwin Scientific CENT BTL 750ML PP BECKMAN 6/PK
SDP
Catalog No. NC9756153
Encompass
The translucent polypropylene bottle is perfect for use in benchtop centrifuges, microcentrifuges rotors and high performance/capacity rotors with specific required adapters. This product has a volume capacity of 750 mL and dimensions of 96 x 130 mm. This is sold in a pack of 6.
Catalog No. NC9756153 Supplier Andwin Scientific Supplier No. BK356855
The translucent polypropylene bottle is perfect for use in benchtop centrifuges, microcentrifuges rotors and high performance/capacity rotors with specific required adapters. This product has a volume capacity of 750 mL and dimensions of 96 x 130 mm. This is sold in a pack of 6.