Learn More
Chrom Tech Inc Centrifuge filter tube, MAXI, CA 0.45um/50PK
SDP
Supplier:  Chrom Tech Inc CTFCA04503
Encompass
Centrifuge filters are very useful for small volume sample filtration. Designed for use in centrifuges, the filter units consist of a sample chamber and a receiver tube. Various membranes are available for bacteria removal, particle removal, HPLC sample preparation, removal of cells from media and DNA removal from agarose or acrylamide gels. The tube has a marking area and is graduated. 25 mL sample capacity. 50 mL graduated receiver tube and cap - Maximum G-force: 2,500xG -Polypropylene housing - Use with Fixed-angle Rotor Centrifuge - Hold up Volume: less than 20 µL - Max Operating Temp: 50 C.
Catalog No. NC1122688