Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foxx Life Sciences EZBio MTO Centrifuge Tube Assembly, 50mL, Polypropylene (PP) Centrifuge Tube, Sampling Tube, Single Use Technology with AdvantaFlex Tubing and Pinchpipe, Vented with Diptube, CPC Aseptiquik (AQS), Sterile, 10/CS
GREENER_CHOICE

Catalog No. 503029718
Encompass_Preferred

The sterile EZBio Centrifuge Tube Assembly is a single use closed loop system designed for aseptic sampling and sterile transfer.
All components are USP Class VI resin, assembled in a Class 7 cleanroom, and gamma-irradiated in the USA. These assemblies feature a closed cap, an 18” inlet line (1/8” x 1/4” TPE tubing) with a pinch clamp, pinch pipe, plug or luer or CPC MPC/AQS, a 1.5” vent line with PTFE hydrophobic filter, and a dip tube. A centrifuge tube stand securely holds either size. The polypropylene resin meets FDA and USP standards.

Features
Designed for contamination-free sampling/transfer within closed systems
Leak-proof seal with a platinum-cured silicone gasket
Available in 15mL and 50mL with six TPE tubing options and four fitting types.
Withstands up to 4 psi; pinch pipe enables quick aseptic separation

Applications
Cell Harvest & Downstream Processing
Drug Substance Purification
Drug Product Formulation
Buffer & Media Preparation

Catalog No. 50-302-9718 Supplier Foxx Life Sciences Supplier No. 58530A3FLS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The sterile EZBio Centrifuge Tube Assembly is a single use closed loop system designed for aseptic sampling and sterile transfer.
All components are USP Class VI resin, assembled in a Class 7 cleanroom, and gamma-irradiated in the USA. These assemblies feature a closed cap, an 18” inlet line (1/8” x 1/4” TPE tubing) with a pinch clamp, pinch pipe, plug or luer or CPC MPC/AQS, a 1.5” vent line with PTFE hydrophobic filter, and a dip tube. A centrifuge tube stand securely holds either size. The polypropylene resin meets FDA and USP standards.

Features
Designed for contamination-free sampling/transfer within closed systems
Leak-proof seal with a platinum-cured silicone gasket
Available in 15mL and 50mL with six TPE tubing options and four fitting types.
Withstands up to 4 psi; pinch pipe enables quick aseptic separation

Applications
Cell Harvest & Downstream Processing
Drug Substance Purification
Drug Product Formulation
Buffer & Media Preparation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.