Learn More
CELLTREAT Scientific Products 25mL Centrifuge Tube, Conical Bottom, Bag, Sterile
SDP
Catalog No. NC3377658
Encompass_Preferred
Convenient volume option between 15mL and 50mL Centrifuge TubesSmaller tube saves storage space and reduces plastic wasteSame diameter as standard 50mL tubes – use the same racks and centrifuge rotorsReduce risk of contamination and increase centrifugation stability with shorter tube
Catalog No. NC3377658 Supplier CELLTREAT Scientific Products Supplier No. 229487
Convenient volume option between 15mL and 50mL Centrifuge TubesSmaller tube saves storage space and reduces plastic wasteSame diameter as standard 50mL tubes – use the same racks and centrifuge rotorsReduce risk of contamination and increase centrifugation stability with shorter tube