Learn More
Caplugs Evergreen 30 mL Centrifuge Tube, Polypropylene, Natural, Screw Cap, Not Sterile, 1000/cs
Catalog No. NC1876226
Encompass_Preferred
This 30 mL centrifuge tube from Evergreen is ideal for general purpose laboratory work and shipping of infectious specimens due to the leakproof caps.
Catalog No. NC1876226 Supplier Caplugs Supplier No. 2403530G30
This 30 mL centrifuge tube from Evergreen is ideal for general purpose laboratory work and shipping of infectious specimens due to the leakproof caps.