Learn More
CELLTREAT Scientific Products 30mL Centrifuge Tube, Self-Standing, Sterile
SDP
Catalog No. NC2131746
Encompass_Preferred
30mL size is an ideal option for intermediate centrifuge tube volumes. Self-standing base for use without a rack. Molded graduations ensure volume accuracy to +/- 2%. Conical bottom for maximized sample retrieval. Easy to write on flat surface caps feature integrated leak-proof seal. Convenient zip-closure bag is a practical alternative to folding and taping opened bags.
This item is not returnable. View return policy
Catalog No. NC2131746 Supplier CELLTREAT Scientific Products Supplier No. 229494
30mL size is an ideal option for intermediate centrifuge tube volumes. Self-standing base for use without a rack. Molded graduations ensure volume accuracy to +/- 2%. Conical bottom for maximized sample retrieval. Easy to write on flat surface caps feature integrated leak-proof seal. Convenient zip-closure bag is a practical alternative to folding and taping opened bags.