Learn More
CELLTREAT Scientific Products 500mL Conical Bottom Centrifuge Tube, Sterile
SDP
Catalog No. NC1964644
Encompass_Preferred
For applications requiring large volume centrifugation. Tubes include molded graduation lines. Polyethylene (HDPE) cap with integrated plug seal provides a secure closure. Temperature range of -80°C to 120°C. Use of a rotor cushion is advised to support tubes during centrifugation.
Catalog No. NC1964644 Supplier CELLTREAT Scientific Products Supplier No. 229463
For applications requiring large volume centrifugation. Tubes include molded graduation lines. Polyethylene (HDPE) cap with integrated plug seal provides a secure closure. Temperature range of -80°C to 120°C. Use of a rotor cushion is advised to support tubes during centrifugation.