Learn More
CELLTREAT Scientific Products 50mL Centrifuge Tube, Metal Free - Bag, Sterile
SDP
Catalog No. NC3377685
Encompass_Preferred
Validated to contain less than 1ppb of more than 30 trace metal elements, ensuring samples are free of contaminantsGraduations are printed in bold, easy-to-read black ink and are accurate to +/-2%Large white marking area provides excellent contrast for identificationEasy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof sealTubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantificationConvenient zip-closure bag is a practical alternative to folding and taping opened bagsEasily refill reusable racks with tubes packed in bags
Catalog No. NC3377685 Supplier CELLTREAT Scientific Products Supplier No. 230482
Validated to contain less than 1ppb of more than 30 trace metal elements, ensuring samples are free of contaminantsGraduations are printed in bold, easy-to-read black ink and are accurate to +/-2%Large white marking area provides excellent contrast for identificationEasy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof sealTubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantificationConvenient zip-closure bag is a practical alternative to folding and taping opened bagsEasily refill reusable racks with tubes packed in bags