Learn More
Andwin Scientific CENT TUBE 50ML PP NS 500/CS
SDP
Catalog No. NC9688848
Encompass_Preferred
The 50 mL SuperClear™ Ultra-High Performance Centrifuge Tubes feature a thick-wall construction that permits high-speed operation. The tubes are also leakproof, providing a secure storage for samples. This product is free of RNase, DNase, and endotox
This item is not returnable. View return policy
Catalog No. NC9688848 Supplier Andwin Scientific Supplier No. 21008240
The 50 mL SuperClear™ Ultra-High Performance Centrifuge Tubes feature a thick-wall construction that permits high-speed operation. The tubes are also leakproof, providing a secure storage for samples. This product is free of RNase, DNase, and endotox