Learn More
CELLTREAT Scientific Products TUBE & CAP, 50mL Centrifuge Tube & Cap - Non-sterile (Caps and Tubes Packed Separately, Bags of 50)
SDP
Catalog No. NC3377686
Encompass_Preferred
Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%Large white marking area provides excellent contrast for identificationEasy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof sealTubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantificationConvenient zip-closure bag is a practical alternative to folding and taping opened bagsPaperboard rack is a recyclable, fold-flat alternative to bulky foam racks
Catalog No. NC3377686 Supplier CELLTREAT Scientific Products Supplier No. 230423
Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%Large white marking area provides excellent contrast for identificationEasy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof sealTubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantificationConvenient zip-closure bag is a practical alternative to folding and taping opened bagsPaperboard rack is a recyclable, fold-flat alternative to bulky foam racks