Learn More
CELLTREAT Scientific Products TUBE ONLY, 50mL Centrifuge Tube, Sterile
SDP
Catalog No. NC3377733
Encompass_Preferred
Manufactured and packaged using an improved process, while maintaining standardsEasy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof sealGraduations are printed in bold, easy-to-read black ink and are accurate to +/-2%Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantificationLarge white marking area provides excellent contrast for identificationConvenient zip-closure bag is a practical alternative to folding and taping opened bags
Catalog No. NC3377733 Supplier CELLTREAT Scientific Products Supplier No. 230428
Manufactured and packaged using an improved process, while maintaining standardsEasy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof sealGraduations are printed in bold, easy-to-read black ink and are accurate to +/-2%Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantificationLarge white marking area provides excellent contrast for identificationConvenient zip-closure bag is a practical alternative to folding and taping opened bags