Learn More
CELLTREAT Scientific Products 10mL Centrifuge Tube - Bag, Sterile
SDP
Catalog No. NC1964632
Encompass_Preferred
Offers the convenience of a micro centrifuge tube with the ability to process sample volumes up to 10mL. Easily access cell pellets and reduce contamination risk with shorter tubes. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small-volume quantification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags.
Catalog No. NC1964632 Supplier CELLTREAT Scientific Products Supplier No. 229493
Offers the convenience of a micro centrifuge tube with the ability to process sample volumes up to 10mL. Easily access cell pellets and reduce contamination risk with shorter tubes. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small-volume quantification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags.