Learn More
Stellar Scientific Conical Centrifuge Tubes 50ml, RNase/DNase Free, STERILE, Foam Racked, 500/cs
SDP
Catalog No. NC1899498
Encompass_Preferred
Our tubes are made from tough polypropylene and are rugged enough to withstand freezing to minus 80 and centrifugal forces up to 15,000 x g. All centrifuge tubes are certified RNase, DNase and pyrogen free. When sterile, they have been gamma irradiated.
Catalog No. NC1899498 Supplier Stellar Scientific Supplier No. T50100
Our tubes are made from tough polypropylene and are rugged enough to withstand freezing to minus 80 and centrifugal forces up to 15,000 x g. All centrifuge tubes are certified RNase, DNase and pyrogen free. When sterile, they have been gamma irradiated.